Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Sanitary Fitting Precision Pump Tubing Assembly, PharMed BPT, L/S 16; 10 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9610316
Encompass
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige Requires sanitary gaskets for both connections to ensure quality seal
Catalog No. NC1290195