Learn More
TARGETMOL CHEMICALS INC Mastoparan acetate72093-21-1 f
SDP
Supplier:  TARGETMOL CHEMICALS INC TP1386L15MG
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Mastoparan acetate is a peptide toxin from wasp venom. It has the chemical structure Ile-Asn-Leu-Lys-Ala-Leu-Ala-Ala-Leu-Ala-Lys-Lys-Ile-Leu-NH2. Purity 100%
Catalog No. 50-287-7322