Learn More
Medchemexpress LLC Mastoparan X | 72093-22-2 | 98.7% | 1555.97 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP27055MG
Encompass_Preferred
Mastoparan X is a GTP-binding regulatory protein (G protein)-activating peptide, and a tetradecapeptide derived from wasp venom. It functions by directly activating G proteins that couple to phospholipase C, leading to secretion from various types of cells. This product is for research use only and not sold to patients.
  • Activates GTP-binding regulatory proteins (G proteins).
  • Derived from wasp venom as a tetradecapeptide.
  • Directly activates G proteins that couple to phospholipase C.
  • Induces secretion from various cell types.
  • Interacts with phospholipid bicelles.
  • Increases cell permeability to cations.
  • Disrupts electrolyte balance and causes cell lysis.
Catalog No. 50-004-00389