Learn More
Amsbio LLC MatriMix (511) Fully Definied 3D ECM Hydrogel for Cell Culture
Supplier:  Amsbio LLC AMS.899 001
Encompass
MatriMix is a 3D culture substrate product developed with the aim of culturing cells in an environment similar to that in vivo. It is fully defined, containing collagen, laminin E8 fragments and hyaluronic acid, and types and concentrations of individual components can be customized to provide the optimal extracellular environment for target cells. MAIN FEATURES: Fully defined | Regulatory friendly (medical grade collagen and laminin is available) | Solution at 4C – becomes a clear gel when incubated at 37C | Multi-organ organoid formation | Suitable for Xenograft transplantation | High batch-to-batch reproducibility between vials | Highly versatile (can be used with a variety of cells including iPSCs and ASCs) | Cell culture of various organ tissues possible by adjusting collagen type and concentration, laminin isoform, and various ECM components | Drug Screening more representative of in vivo testing compared to Matrigel™ and other BME equivalents.
Catalog No. 50-256-6190