Learn More
Essendant MATWATERGUARD 3X5BR
Supplier:  Essendant MLLWG030514
Encompass_Preferred
These mats are recommended as an indoor mat but substitutes as an outdoor mat in warm to moderate climates The bi-level construction allows dirt and debris to get trapped below the walking surface and off your floors The polypropylene carpeted surface makes this a scraper to remove debris and also a wiper to help trap moisture The non-slip all-rubber cleated backing provides a secure gripping action to carpets and hard surface floors The beveled edge border helps contain the dirt and moisture and helps reduce the chance of slips trips and falls Easy-to-clean and maintain just shake out vacuum hose and dry or clean with a light carpet cleaning machine Mat Type- Wiper Mat Application- Indoor Width- 36 Length- 60
Catalog No. 50-000-33774