Learn More
Novus Biologicals Maurice CE-SDS Molecular Weight Markers
SDP
Supplier:  Novus Biologicals 046432
Encompass
Lyophilized molecular weight markers (10, 20, 33, 55, 103, 178 & 270 kDa) for 8 runs.
Catalog No. NC1306507