Learn More
Novus Biologicals Maurice CE-SDS Wash Solution, 20mL
SDP
Supplier:  Novus Biologicals 046569
Encompass
Wash solution for CE-SDS application, 20 mL. For use with CE-SDS cartridge (PS-MC02-S) on Maurice and Maurice S. systems only.
Catalog No. NC1642866