Learn More
Novus Biologicals Maurice cIEF pI Marker - 9.99, Lyophilized, 210 uL
SDP
Supplier:  Novus Biologicals 046034
Encompass
Maurice C. reagents and consumables are designed for use on with Maurice and Maurice C. systems, and must be used in conjunction with Compass for iCE version 1.0 software.
Catalog No. NC1227077