Learn More
Medchemexpress LLC HY-109037 1mg , Mavacamten CAS:1642288-47-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090371MG
Encompass_Preferred
HY-109037 1mg Mavacamten CAS: 1642288-47-8 Mavacamten (MYK461) is an orally active modulator of cardiac myosin, with IC50s of 490, 711 nM for bovine cardiac and human cardiac, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0499