Learn More
Medchemexpress LLC Mavacamten-d7 | MFCD31746877 | 98.0% | 280.37 g/mol | C15H12D7N3O2 | 5 MG

Supplier: Medchemexpress LLC HY109037S45MG
Mavacamten-d7 is the deuterium-labeled (d7) stable isotope of mavacamten provided as an analytical reference for use in mass spectrometry, isotope-dilution assays, and related research applications. It is intended as a reference standard for quantitation and method development and is characterized by its labeled hydrogen atoms and specified molecular properties.
- Deuterium-labeled for use as an internal standard in analytical methods.
- Suitable for mass spectrometry and isotope-dilution assays.
- Molecular formula: C15H12D7N3O2.
- Molecular weight: 280.37 g/mol.
- Supplied in a small reference quantity (5 mg) for laboratory use.
- Intended for research and analytical applications, not for human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.