Learn More
TARGETMOL CHEMICALS INC Mavatrep 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T160145MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Mavatrep (JNJ-39439335) is a selective antagonist of TRPV1 with Ki of 6.5 nM and can be used for studies about inflammatory pain. Purity 98.38%
Catalog No. 50-287-6069