Learn More
BMEM BIO Max Pluripotent
Supplier:  BMEM BIO 1004
Encompass
BMEM Max Pluripotent is a highly optimized, chemically defined, and animal product-free medium and supplement combination for the maintenance of human iPSCs.•Beginner Friendly Formulation: BMEM Max Basal Medium is engineered for enhanced cell survival under suboptimal conditions, making it ideal for single-cell cloning, CRISPR modification, and overgrowth scenarios.•Easy Adaptation: The Primed Supplement resolves adaptation issues from legacy media and features enhanced stability (room temperature shipment).•Flexible Protocols: Validated for weekend-free and no-change feeding schedules.•Matrix Compatibility: Compatible with standard matrices; however, use with animal component-free matrices (e.g., Laminin-521, Synthemax) is highly recommended.•Versatile Passaging: Works with Accutase, TrypLE™, ReLeSR™, and Versene.•Quality: Performance tested on hiPSCs and sterility tested.
Catalog No. NC3932633