Learn More
Active Motif MAXbind™ Staining Medium
SDP
Supplier:  Active Motif 15253
Encompass
MAXbind Staining Medium is a non-mammalian incubation agent that optimizes antibody staining in immunofluorescence and immunostaining assays. Used with MAXblock Blocking Medium, MAXbind promotes superior antibody binding while eliminating non-specific primary and secondary antibody binding. MAXbind has proven effective in immunofluorescence, immunohistochemistry, Western blotting and ELISA.
Catalog No. NC1841523