Learn More
IBI SCIENTIFIC 3P MAXI FAST-ION PLSMD KT-25 PREP
Supplier:  IBI SCIENTIFIC 3P IB47122
Encompass_Preferred
DNA Purification Kit, Fast-Ion Maxi Plasmid Kit (25-Prep), IBI Scientific, Sample Size: 100ml for high copy plasmid & 250ml for low copy plasmid, Binding Capacity: 1mg, Operation Time: 120min, Expectant Yield: 500ug-1mg of plasmid DNA.
Catalog No. NC0710909