Learn More
Enzymax LLC Maxi Filter column for lysate clarification
SDP
Supplier:  Enzymax LLC EZC111F
Encompass_Preferred
Maxi Filter columns are designed to work with DNA or RNA maxi columns. It is used for clarification of lysate during DNA/RNA maxi preparation with maxi DNA/RNA binding columns (either silica based or ion-exchange column).
Catalog No. NC1814925