Learn More
Waters Corp Waters Corporation UPLC Method Development Kit, Maximum Selectivity RP/HILIC, 1.7 μm
Supplier:  Waters Corp 176002127
Encompass_Preferred
The Waters Corporation Maximum Selectivity RP and HILIC UPLC Method Development Kit streamlines method development for UPLC and UHPLC systems. It includes a variety of column chemistries for reversed-phase and HILIC separations.
  • Features low silanol activity.
  • Includes eCord technology.
  • Contains CSH C18, CSH Phenyl-Hexyl, CSH Fluoro-Phenyl, and BEH Amide chemistries.
  • Designed for reversed-phase and HILIC separations.
Catalog No. 50-791-101
Add to Cart
Request a Quote