Learn More
Fluidigm Corp MAXPAR ANTIBODY LABELING KIT
SDP
Supplier:  Fluidigm Corp 201153A
Encompass
This Maxpar kit lets you conjugate IgG antibodies with the 153Eu metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.
Catalog No. NC1221484