Learn More
Electron Microscopy Sciences May-Grunwald Stain Solution 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2606405
Encompass_Preferred
May-Grunwald Staining solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is used for bone marrow staining.
Catalog No. 50-270-51