Learn More
ATCC ATCC Mayaro Virus (VR-1966), strain 07-18066-99 - frozen BSL-2 viral stock; propagate in Vero 76 cells
Supplier:  ATCC VR1966
Encompass
Mayaro virus (ATCC VR-1966) is an Alphavirus research strain supplied as frozen viral stock for laboratory research. The isolate was recovered from human serum in Tingo María, Huánuco, Peru. The product is designated biosafety level 2 and is intended for research use; propagation is performed in Vero 76 cells.
  • Frozen viral stock for storage and transport.
  • Isolated from human serum in Tingo María, Huánuco, Peru.
  • Propagates in Vero 76 cells.
  • Designated biosafety level 2; handle under appropriate containment.
  • Suitable for virology and pathogenesis research applications.
  • Genome sequence available from the provider's genome portal.
Catalog No. NC3665010