Learn More
ATCC ATCC Mayaro Virus Control Ascitic Fluid (VR-1277CAF)
Supplier:  ATCC VR1277CAF
Encompass
Freeze-dried ascitic fluid control material for Mayaro virus used as a quality control in virology assays. Intended for laboratory research use only; not for diagnostic, therapeutic, or consumptive use. Commercial use requires a license from the manufacturer.
  • Freeze-dried ascitic fluid format supplied as a control material.
  • Intended for laboratory research use only; not for diagnostic or therapeutic use.
  • Biosafety level 1 handling recommended.
  • Viability warranted for 30 days from shipment when stored per manufacturer instructions.
  • Import permits or documentation may be required for certain jurisdictions.
Catalog No. NC3665017