Learn More
Electron Microscopy Sciences Mayer's Hematoxylin 4 LT
SDP
Supplier:  Electron Microscopy Sciences 2604310
Encompass_Preferred
Mayer's Hematoxylin solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It stains selectivity for nuclei.
Catalog No. 50-270-49