Learn More
Electron Microscopy Sciences Mayers Hematoxylin 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2630601
Encompass_Preferred
Used in Laqueur's Method for Alcoholic Hyalin for mallory bodies, erythrocytes, bile pigment and proteinaceous, material in liver.
Laqueur, G.I., Amer. J. Clin. Path., 20:689-690, 1950, Luna, L.G. (ed), Manual of Hitopathologic Staining Method of the AFIP, 3rd ed, McGraw-Hill, NY, c. 1968, p 160
Lillie, R.D., Fullmer, H.M., Histopathologic Technic and Practical Histochemistry 4th ed., cGraw-Hill, NY, 1976, p 670-671
Clark, G., (ed) Staining Procedures, 3rd ed. WilliamS & Wilkins, Baltimore, p 51, c. 1973
Catalog No. 50-317-41