Learn More
Electron Microscopy Sciences Mayers Mucihematein, Modified 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2632101
Encompass_Preferred
Used in Modification of Mayer's Mucihematein for staining mucins, especially those derived from epithelial cells of glandular tissue. See Laskey, Alice: Stain Technology, 25:33 (1959) or Clark G.: Staining Procedures, Williams & Wilkins Co., Baltimore, 3rd Ed., c. 1973, p. 57.
Catalog No. 50-317-50