Learn More
Med Vet International Miltex Vantage Mayo-Hegar Needle Holder, 6in
Supplier: Med Vet International MILV9844
The Miltex Vantage V98-44 Mayo-Hegar needle holder is made from high quality Japanese stainless steel and measures 6" in length. It has a finger ring handle with a ratchet lock and straight, serrated jaws.