Learn More
Electron Microscopy Sciences EMS SuperCut Mayo Scissors with Curved Blades, 9"
SDP
Supplier:  Electron Microscopy Sciences 7299616
Encompass_Preferred
SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings Curved version cuts into heavier or deeper tissue.
Blade: Curved
OAL: 9"
Catalog No. 50-193-1423