Learn More
Sklar Instruments Mayo Tray, Sklar, 10 in. x 6-1/2 in. x 3/4 in. Mayo Tray Perforated, Stainless Steel. The Mayo Tray holds instruments to be used in a surgical procedure. EA/1.
SDP
Supplier:  Sklar Instruments 101684
Encompass_Preferred
Mayo Tray, Sklar, 10 in. x 6-1/2 in. x 3/4 in. Mayo Tray Perforated, Stainless Steel. The Mayo Tray holds instruments to be used in a surgical procedure. It is typically organized and monitored by the scrub person or surgical technologist assisting in the procedure. It is used in conjunction with a stand so that the tray and instruments are situated above the waist and easily accessible during the procedure. The tray holds instruments that are the most frequently used during any given procedure. The tray is available perforated or non perforated and in a variety of sizes. It is sold individually, in packs of 6, and packs of 12.
Catalog No. 50-143-7092