Learn More
Medchemexpress LLC HY-116624 25mg Medchemexpress, MAZ51 CAS:163655-37-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11662425MG
Encompass_Preferred
Medchemexpress, HY-116624 25mg MAZ51 CAS:163655-37-6 MAZ51 is a selective inhibitor of VEGFR-3 (Flt-4) tyrosine kinase. MAZ51 inhibits VEGF-C-induced activation of VEGFR-3 without blocking VEGF-C-mediated stimulation of VEGFR2. MAZ51 had no effect on ligand-induced autophosphorylation of EGFR, IGF-1R and PDGFRβ. MAZ51 blocks proliferation and induces apoptosis in a wide variety of tumor cells. Antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9433