Learn More
Medchemexpress LLC Mazdutide | 2259884-03-0 | 99.9% | 4563.06 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP33751MG
Encompass_Preferred
Mazdutide (IBI-362) is a long-acting synthetic oxyntomodulin analog that acts as a co-agonist of glucagon-like peptide (GLP-1R) and glucagon receptor (GCGR). It stimulates insulin secretion and is utilized in studies concerning obesity and type 2 diabetes.
  • Long-acting synthetic oxyntomodulin analog
  • Co-agonist of GLP-1R and GCGR
  • Binds to human and mouse GCGR (Ki: 17.7 nM and 15.9 nM, respectively)
  • Binds to GLP-1R (Ki: 28.6 nM and 25.1 nM, respectively)
  • Stimulates insulin secretion from mouse islets (EC50: 5.2 nM)
  • Reduces body weight and physical intake in diabetic mice with diet-induced obesity
Catalog No. 50-003-99283