Learn More
Medchemexpress LLC MBL1 Rat HEK293 F 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP770795UG
Encompass_Preferred
MBL1 protein is a calcium-dependent lectin that participates in innate immunity by binding to mannose fucose and N-acetylglucosamine on microorganisms and activating the lectin complement pathway MBL1 Protein Rat (HEK293 Fc) is the recombinant rat-derived MBL1 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-85702