Learn More
Genetex Inc Myelin basic protein Monoclonal Antibody [MSVA-390R] HistoMAX™ (GTX04435) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04435
Encompass_Preferred
The Myelin basic protein antibody [MSVA-390R] HistoMAX™ (GTX04435) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human cerebrum (grey matter) tissue using GTX04435 Myelin basic protein antibody [MSVA-390R] HistoMAX. In the cerebrum a fibrillar myelin basic protein immunostaining occurs along intracerebral axons. Neurons are not stained.
Catalog No. NC3666944