Learn More
Research Products International Corp MC-24 Touch Screen High Speed MicroCentrifuge
SDP
Supplier:  Research Products International Corp 248580
Encompass
MC-24 Touch Screen MicroCentrifuge features compact size with small footprint for labs requiring a smart unit that performs molecular biology applications in a snap. Unit features large, full color touch screen that shows actual speed (RPM or RCF) with remaining time; stores up to 9 speed/time profiles; mySpin, 1 button program recall; unique air-flow system keeps samples cool during spin cycles; momentary spin; and quiet operation. Supplied with Combi-Rotor that accepts 24 x 1.5- 2.0ml and 2 x 8 strips of 0.2ml PCR tubes and includes Quick-Seal lid that opens and locks with 1/4 turn.
Full color touch screen
COMBI-Rotor accepts tubes and strips
Compact size
Two year warranty
Catalog No. 50-222-9740