Learn More
Medchemexpress LLC MC-Ala-Ala-PAB | 1949793-44-5 | 97.3% | 458.51 g/mol | C23H30N4O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14846450MG
Encompass_Preferred
MC-Ala-Ala-PAB is a cleavable antibody-drug conjugate (ADC) linker supplied as an Fmoc-protected building block. Fmoc-Ala-Ala-PAB contains an Ala-Ala dipeptide spacer and a para-aminobenzyl (PAB) self-immolative spacer and is used in ADC synthesis and conjugation chemistry.
  • Cleavable dipeptide linker suitable for ADC development.
  • Fmoc-protected for standard solid-phase and solution-phase chemistry.
  • High purity (97.3%) for research applications.
  • Available in multiple pack sizes, including 50 mg.
  • Recommended storage: powder at -20°C for long-term stability.
Catalog No. 50-002-02101