Learn More
Medchemexpress LLC MC-(β-Ala)-PABC-(β-D-GlcUA)-amide-PEG1-CH2-CC-885 | 2722697-86-9 | 99.6% | 1133.55 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1604971MG
Encompass_Preferred
This compound is a neodegrader conjugate that can be utilized in the synthesis of antibody neoDegrader conjugates (AnDC). It is intended for research purposes only and is not sold to patients.
  • This conjugate is useful for the synthesis of antibody neodegrader conjugates.
  • It is available as a solid with a white to off-white color.
  • The recommended shipping condition is room temperature within the continental US, with potential variations elsewhere.
  • For storage, it should be kept at -20°C in sealed storage, away from moisture and light.
  • When dissolved in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, under sealed storage away from moisture and light.
  • Its solubility in DMSO is 100 mg/mL, requiring sonication.
Catalog No. 50-003-91420