Learn More
Medchemexpress LLC Mc-Alanyl-Alanyl-Asparagine-PAB-MMAE | 1638970-46-3 | 99.8% | 1316.58 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1472041MG
Encompass_Preferred
Mc-Alanyl-Alanyl-Asparagine-PAB-MMAE is a potent anticancer agent specifically activated by the tumor microenvironment. It effectively suppresses tumor growth in mice, as demonstrated by in vivo studies where it reduced Panc-1 tumor growth when administered intravenously weekly for 4 weeks.
  • Potent anticancer agent
  • Specifically activated by the tumor microenvironment
  • Suppresses tumor growth in mice
  • Demonstrated efficacy against Panc-1 tumors in vivo
  • Provided as a white to off-white solid
  • High purity, 99.8%
  • Soluble in DMSO (180 mg/mL)
Catalog No. 50-003-62858