Learn More
YMC America, Inc MC C8 80A 2.7um 30x2.1mm
SDP
Supplier:  YMC America, Inc COS08SQ703Q1WT
Encompass
Meteoric Core columns offer outstanding resolution for UHPLC and HPLC with a base particle of core-shell type silica gel Meteoric Core can be used over a wide pH range providing excellent peak shape for basic and coordination compounds When compared to conventional fully-porous materials under the same conditions Meteoric Core columns can shorten analysis times by two thirds Theoretical plate count is maintained at 2X faster flow rate while resolution is maintained at an operating pressure below 5 000 psi Meteoric Core is available in three phases including C18 widepore C18 BIO and C8 for medium hydrophobicity These prepacked analytical columns are ideal for ultra-fast and high-resolution analyses on both HPLC and UHPLC systems
Catalog No. 50-008-79924