Learn More
Medchemexpress LLC Mc-d-val-cit-pab-pnp | 1350456-66-4 | 98.1% | 737.76 | C35H43N7O11 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY147021100MG
Encompass_Preferred
MC-Val-D-Cit-PAB-PNP is a cleavable peptide linker reagent used for constructing antibody-drug conjugates (ADCs) and other linker-payload conjugates. It contains functional groups that enable antibody conjugation and payload coupling, and is supplied as a solid with high purity suitable for research use.
  • Cleavable peptide linker suitable for antibody-drug conjugate synthesis.
  • Contains a maleimidocaproyl group for antibody conjugation.
  • Includes a p-nitrophenyl leaving group for payload coupling.
  • Delivered as a white to off-white solid with high purity.
  • Stable as a powder at -20°C for long-term storage; different stability in solution.
Catalog No. 50-002-02664