Learn More
Medchemexpress LLC Mc-d-Val-d-Cit-PAB-PNP | 1350456-68-6 | 98.8% | 737.76 | C35H43N7O11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17316150MG
Encompass_Preferred
Mc-d-Val-d-Cit-PAB-PNP is a cleavable peptide linker used for synthesizing antibody-drug conjugates (ADCs). Supplied as a white to off-white solid with a molecular weight of 737.76 and measured purity of 98.80%, it is intended for research applications and for use in controlled payload release from ADCs.
  • Cleavable peptide linker suitable for antibody-drug conjugate synthesis.
  • High purity for consistent conjugation performance.
  • White to off-white solid, easy to handle in the laboratory.
  • Available in milligram pack sizes for small-scale research use.
  • Store at recommended low temperatures to maintain stability.
Catalog No. 50-002-03314