Learn More
Medchemexpress LLC Mc-d-val-d-cit-pab-pnp | 1350456-68-6 | MFCD22200431 | 98.8% | 737.76 | C35H43N7O11 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1731615MG
Encompass_Preferred
A protease-cleavable peptide linker used for synthesizing antibody-drug conjugates (ADCs). The linker contains a Val-Cit dipeptide and a para-nitrophenyl (PNP) leaving group, providing enzymatic payload release after internalization. Intended for research use only.
  • Cleavable Val-Cit dipeptide linker.
  • Maleimide-compatible conjugation handle for payload attachment.
  • PNP leaving group for facile conjugation chemistry.
  • High purity with analytical data (COA, HNMR, LCMS, RP-HPLC).
  • Available in multiple small pack sizes for research workflows.
Catalog No. 50-002-04075