Learn More
Medchemexpress LLC HY-114233 5mg Medchemexpress, MC-GGFG-DX8951 CAS:1600418-29-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1142335MG
Encompass_Preferred
Medchemexpress, HY-114233 5mg MC-GGFG-DX8951 CAS:1600418-29-8 MC-GGFG-DX8951 is a drug-linker conjugate for ADC with antitumor activity by using DX8951 (a DNA topoisomerase I inhibitor), linked via the protease cleavable MC-GGFG linker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9780