Learn More
Medchemexpress LLC MC-Gly-Gly-Phe-Gly | 2413428-36-9 | 98.0% | 529.54 g/mol | C25H31N5O8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY4424650MG
Encompass_Preferred
MC-Gly-Gly-Phe-Gly is a protease-cleavable peptide linker used for research in antibody-drug conjugate (ADC) construction. It contains a maleimide (MC) functional group and a Gly-Gly-Phe-Gly sequence that enables enzymatic payload release.
  • Cleavable tetrapeptide linker with protease-sensitive Gly-Gly-Phe-Gly sequence.
  • Maleimide functional group enables thiol conjugation to antibodies.
  • Molecular weight 529.54 g/mol; formula C25H31N5O8.
  • High purity suitable for research applications.
  • Available in small research pack sizes (e.g., 50 MG).
Catalog No. 50-002-04100