Learn More
Medchemexpress LLC MC-Gly-Gly-Phe-Gly-PAB-PNP | 2632342-16-4 | 99.2% | 799.78 g/mol | C39H41N7O12 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY14716425MG
Encompass_Preferred
MC-Gly-Gly-Phe-Gly-PAB-PNP is a protease-cleavable tetrapeptide linker used to assemble antibody-drug conjugates (ADCs). It contains a Gly-Gly-Phe-Gly recognition sequence, a para-aminobenzyl self-immolative spacer, and a p-nitrophenyl leaving group that enables controlled payload release.
  • Protease-cleavable GGFG recognition sequence.
  • Para-aminobenzyl self-immolative spacer with p-nitrophenyl leaving group.
  • High chemical purity for research use.
  • Soluble in DMSO for conjugation workflows.
  • Offered in milligram-scale package sizes for lab-scale synthesis.
Catalog No. 50-002-02107