Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MC-VA-PABC-MMAE | 1818864-51-5 | 99.9% | 1230.53 | C65H99N9O14 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1472395MG

Encompass_Preferred

MC-VA-PABC-MMAE is an agent-linker conjugate used in antibody-drug conjugate (ADC) research that links a peptide-based cleavable linker to the cytotoxic payload monomethyl auristatin E (MMAE). The material is supplied as a white to off-white solid for research use only and is intended for in vitro ADC assembly and evaluation.

  • Contains a peptide MC-VA-PABC linker coupled to MMAE.
  • High purity suitable for research applications.
  • Molecular weight approximately 1230.53 Da.
  • Chemical formula C65H99N9O14.
  • Supplied as a solid, white to off-white in appearance.
  • Store sealed at -20°C; in solvent store at -80°C for long-term.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-03253


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.