Learn More
Cayman Chemical Mc-Val-CIt-PAB 10mg
SDP
Supplier:  Cayman Chemical 2828510 MG
Encompass_Preferred
A peptide linker molecule; used in the synthesis of ADCs; has been used in the synthesis of an ADC containing the tubulin polymerization inhibitor KGP05; has been used as a precursor in the synthesis of Mc-Val-Cit-PABC-PNP
Catalog No. 50-293-9203