Learn More
Medchemexpress LLC Mc-Val-Cit-PAB-Cl | 1639351-92-0 | 95.0% | 591.10 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112099
Encompass_Preferred
Mc-Val-Cit-PAB-Cl is a cleavable ADC linker. It can be used to conjugate MMAE and antibody to form antibody-MC-vc-MMAE (e.g., anti-CD22-MC-VC-PABC-MMAE with IC50s of 3.3 and 0.95 nM for BJAB and WSU cell lines in cytotoxicity assay).
  • Cleavable ADC linker
  • Used to conjugate MMAE and antibody
  • Forms antibody-MC-vc-MMAE
  • For research use only
Catalog No. 50-004-38826