Learn More
Medchemexpress LLC Mc-val-cit-pab-cl | 1639351-92-0 | MFCD32201033 | 95.0% | 591.10 | C28H39ClN6O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1120995MG
Encompass_Preferred
Mc-Val-Cit-PAB-Cl is a cleavable antibody-drug conjugate (ADC) linker that combines a valine-citrulline dipeptide with a para-aminobenzyl (PAB) self-immolative spacer to enable protease-triggered release of cytotoxic payloads such as MMAE. Supplied as a solid reagent with defined purity, it is intended for ADC linker synthesis and conjugation workflows; follow recommended low-temperature, inert storage for stability.
  • Cleavable val-cit dipeptide linked to a PAB self-immolative spacer.
  • Designed for protease-triggered release of payloads such as MMAE.
  • Provided as a solid reagent suitable for conjugation reactions.
  • High purity for research-grade synthesis and analytical consistency.
  • Stable when stored under inert atmosphere at recommended low temperatures.
Catalog No. 50-002-64163