Learn More
Medchemexpress LLC Mc-val-cit-pab-clindamycin | 1639793-13-7 | 100.0% | 1016.08 g/mol | C46H72Cl2N8O11S | 5 MG

Supplier: Medchemexpress LLC HY1289075MG
MC-Val-Cit-PAB-clindamycin is a drug-linker conjugate used in antibody-drug conjugate (ADC) research that links the protein-synthesis inhibitor clindamycin to a cleavable Val-Cit-PAB linker, producing a payload module with reported antitumor activity.
- High reported purity: 99.99%.
- Chemical formula: C46H72Cl2N8O11S.
- Molecular weight: 1016.08 g/mol.
- Intended for ADC research and preclinical evaluation.
- Supplied as a 5 mg pack.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.