Learn More
Medchemexpress LLC MC-Val-Cit-PAB-dimethylDNA31 | 1639352-06-9 | 1518.15 | C78H101ClN10O19 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12890510MG
Encompass_Preferred
MC-Val-Cit-PAB-dimethylDNA31 is an agent-linker conjugate used in antibody-drug conjugate (ADC) research that links a cleavable valine-citrulline PAB linker to the dimethylDNA31 payload for preclinical conjugation and efficacy studies.
  • Agent-linker conjugate for ADC research and conjugation chemistry.
  • Cleavable valine-citrulline p-aminobenzyl (PAB) linker enables intracellular payload release.
  • DimethylDNA31 payload with reported antitumor and bactericidal activity against persisters.
  • Available in small-scale pack sizes suitable for preclinical studies.
  • Storage: -20°C sealed; in solvent -80°C for long-term stability.
Catalog No. 50-002-02337