Learn More
Medchemexpress LLC MC-Val-Cit-PAB-dimethylDNA31 | 1639352-06-9 | 1518.15 g/mol | C78H101ClN10O19 | 5 MG

Supplier: Medchemexpress LLC HY1289055MG
MC-Val-Cit-PAB-dimethylDNA31 is a conjugate of the protease-cleavable MC-Val-Cit-PAB linker and the dimethylDNA31 payload, developed for antibody-drug conjugate research, conjugation workflows, and preclinical in vitro studies.
- Uses a protease-cleavable Val-Cit-PAB linker for controlled payload release.
- Intended for ADC research, conjugation, and preclinical in vitro studies.
- Available pack sizes: 1 mg, 5 mg, 10 mg.
- Molecular weight: 1518.15 g/mol.
- Molecular formula: C78H101ClN10O19; CAS number: 1639352-06-9.
- Store at -20°C sealed; in solvent: -80°C up to 6 months, -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.