Learn More
Medchemexpress LLC Mc-val-cit-pab-doxorubicin | 159857-70-2 | MFCD32693347 | 99.7% | 1142.17 | C56H67N7O19 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW19092725MG
Encompass_Preferred
MC-Val-Cit-Doxorubicin is a drug-linker conjugate used as an ADC payload and precursor in research. It couples doxorubicin to a Val-Cit-PAB cleavable linker and is designed for conjugation to thiol-containing molecules via a maleimide, enabling enzymatic release of the cytotoxic payload in target cells.
  • Val-Cit cleavable peptide linker enables enzymatic payload release.
  • Maleimide functionality enables conjugation to thiol-containing biomolecules.
  • Includes doxorubicin as the cytotoxic payload for ADC research.
  • High purity 99.7% suitable for analytical and synthetic workflows.
  • Available in multiple small-scale sizes for research needs.
Catalog No. 50-002-81804