Learn More
Medchemexpress LLC MC-Val-Cit-PAB-MMAE | 646502-53-6 | MFCD25372036 | 1316.63 | C68H105N11O15 | 10 MG

Supplier: Medchemexpress LLC HY15575G10MG
VcMMAE (MC-Val-Cit-PAB-MMAE) (GMP) is a GMP-produced drug-linker conjugate used as the payload-linker component in antibody-drug conjugates (ADCs). The molecule uses a valine-citrulline cleavable linker to enable protease-triggered release of monomethyl auristatin E (MMAE), and it is supplied as a solid intended for research applications. The product is shipped on dry ice and should be stored at -20°C, protected from moisture and light.
- Manufactured under Good Manufacturing Practice (GMP) guidelines.
- Cleavable valine-citrulline linker enables controlled release of MMAE.
- Intended for research use only.
- Supplied as an off-white to light yellow solid.
- Store at -20°C, sealed, protect from moisture and light.
- Shipped on dry ice to maintain stability.
- Molecular weight 1316.63 g/mol.
- Chemical formula C68H105N11O15.
- CAS no. 646502-53-6.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.